The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172301.3 | NP_758505.2 | 4 - 5 | 621 | 122 |
GenBank mRNA | AK136259.1 | - | 4 - 5 | 635 | 122 |
AK143597.1 | - | 4 - 5 | 577 | 122 | |
AK163259.1 | - | 4 - 5 | 634 | 122 | |
AK166686.1 | - | 4 - 5 | 635 | 122 | |
AK167950.1 | - | 4 - 5 | 635 | 122 | |
AK168441.1 | - | 4 - 5 | 635 | 122 | |
AK168502.1 | - | 4 - 5 | 635 | 122 | |
AK168993.1 | - | 4 - 5 | 632 | 122 | |
BC011478.1 | - | 4 - 5 | 621 | 122 | |
BC085238.1 | - | 4 - 5 | 617 | 122 | |
S43105.1 | - | 4 - 5 | 659 | 122 | |
X58708.1 | - | 4 - 5 | 625 | 122 | |
X64713.1 | - | 4 - 5 | 612 | 122 |