Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Nlrp9c
Entrez Gene ID
330490
Gene Name
NLR family, pyrin domain containing 9C
Gene Aliases
AA636537, BB062542, Nalp-zeta, Nalp9c
Chromosome Location
Chr.7: 26364702 - 26403682 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY360472.1 - - 2530 105

View Full Product Details