The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025789.5 | NP_080065.4 | 6 - 7 | 1424 | 73 |
GenBank mRNA | AK007979.1 | - | 6 - 7 | 1057 | 73 |
AK016364.1 | - | 6 - 7 | 1010 | 73 | |
AK148230.1 | - | 6 - 7 | 1386 | 73 | |
BC054439.1 | - | 6 - 7 | 1092 | 73 | |
BC057649.1 | - | 6 - 7 | 1383 | 73 | |
BC085294.1 | - | 6 - 7 | 1072 | 73 | |
BC150663.1 | - | 6 - 7 | 909 | 73 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083945.1 | NP_001077414.1 | 6 - 7 | 1113 | 73 |
GenBank mRNA | AK006083.1 | - | 6 - 7 | 1113 | 73 |
AK016473.1 | - | 6 - 7 | 1071 | 73 | |
AK076872.1 | - | 6 - 7 | 1115 | 73 | |
AK161285.1 | - | 6 - 7 | 1076 | 73 |