Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syt8
Entrez Gene ID
55925
Gene Name
synaptotagmin VIII
Gene Aliases
-
Chromosome Location
Chr.7: 142434462 - 142440399 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285857.1 NP_001272786.1 4 - 5 482 86
NM_001285858.1 NP_001272787.1 4 - 5 493 86
NM_001285861.1 NP_001272790.1 4 - 5 483 86
NM_018802.5 NP_061272.2 4 - 5 604 86
GenBank mRNA AB026805.1 - 4 - 5 483 86
AK075832.1 - 4 - 5 605 86
BC100748.1 - 4 - 5 458 86
BC103602.1 - 4 - 5 458 86
BC103665.1 - 4 - 5 469 86
U20107.1 - 2 - 3 264 86

View Full Product Details