The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016754.5 | NP_058034.1 | 4 - 5 | 429 | 70 |
GenBank mRNA | AK003149.1 | - | 4 - 5 | 334 | 70 |
AK010483.1 | - | 4 - 5 | 430 | 70 | |
BC055869.1 | - | 4 - 5 | 313 | 70 | |
U77943.1 | - | 3 - 4 | 276 | 70 |