The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294162.1 | NP_001281091.1 | 10 - 11 | 2631 | 90 |
NM_013569.2 | NP_038597.2 | 14 - 15 | 3659 | 90 | |
GenBank mRNA | AA145089.1 | - | 1 - 2 | 96 | 90 |
AF012868.1 | - | 14 - 15 | 3665 | 90 | |
AF012869.1 | - | 10 - 11 | 2579 | 90 | |
AF034762.1 | - | 10 - 11 | 2623 | 90 | |
AY374424.1 | - | 14 - 15 | 3344 | 90 | |
BC051233.1 | - | 4 - 5 | 697 | 90 | |
CX565252.1 | - | 3 - 4 | 674 | 90 |