Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prl3c1
Entrez Gene ID
27372
Gene Name
prolactin family 3, subfamily c, member 1
Gene Aliases
PLP-J, Prlpi, Prlpj, decidualin
Chromosome Location
Chr.13: 27196660 - 27203749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163218.1 NP_001156690.1 3 - 4 348 106
NM_013766.2 NP_038794.1 3 - 4 318 106
GenBank mRNA AB019118.1 - 3 - 4 315 106
AF150740.1 - 3 - 4 328 106
AF234637.1 - 3 - 4 318 106
AK014980.1 - 3 - 4 348 106
BC120576.1 - 3 - 4 306 106
BC120578.1 - 3 - 4 306 106

View Full Product Details