Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uaca
Entrez Gene ID
72565
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
2700059D02Rik, nucling
Chromosome Location
Chr.9: 60793206 - 60880370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028283.2 NP_082559.1 16 - 17 4008 74
GenBank mRNA AB030647.1 - 16 - 17 4078 74
AK012454.1 - 1 - 2 1064 74
AK173203.1 - 4 - 5 4080 74
BC033470.1 - 17 - 18 3415 74
BC042415.1 - 16 - 17 4008 74

View Full Product Details