Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Izumo4
Entrez Gene ID
71564
Gene Name
IZUMO family member 4
Gene Aliases
9030607L17Rik, AI550407
Chromosome Location
Chr.10: 80702199 - 80705382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027829.3 NP_082105.3 4 - 5 440 72
GenBank mRNA BF682661.1 - 4 - 5 375 72

View Full Product Details