Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Izumo4
Entrez Gene ID
71564
Gene Name
IZUMO family member 4
Gene Aliases
9030607L17Rik, AI550407
Chromosome Location
Chr.10: 80702199 - 80705382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027829.3 NP_082105.3 9 - 10 616 74
GenBank mRNA BF682661.1 - 9 - 10 550 74
CB055593.1 - 8 - 9 616 74

View Full Product Details