The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009759.4 | NP_033889.2 | 5 - 6 | 601 | 56 |
GenBank mRNA | AF012104.1 | - | 4 - 5 | 454 | 56 |
AK036707.1 | - | 4 - 5 | 460 | 56 | |
AK040936.1 | - | 5 - 6 | 602 | 56 | |
AK080038.1 | - | 5 - 6 | 602 | 56 | |
BC138104.1 | - | 5 - 6 | 478 | 56 | |
BC138105.1 | - | 5 - 6 | 478 | 56 | |
U88091.1 | - | 5 - 6 | 587 | 56 |