Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sdc2
- Entrez Gene ID
-
15529
- Gene Name
- syndecan 2
- Gene Aliases
-
4833414L08Rik, AA960457, Hspg1, Synd2, syndecan-2
- Chromosome Location
-
Chr.15: 32920723 - 33034721 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC003929.1 |
- |
2 - 3 |
231 |
77 |
View Full Product Details