Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr13
Entrez Gene ID
18310
Gene Name
olfactory receptor 13
Gene Aliases
K7, MOR261-6
Chromosome Location
Chr.6: 43173988 - 43174920 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146652.1 NP_666863.1 1 - 1 882 125
GenBank mRNA BC120850.1 - 1 - 1 918 125

View Full Product Details