Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4galnt4
Entrez Gene ID
330671
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 4
Gene Aliases
BC038881, Gm1095, beta4GalNAcT4
Chromosome Location
Chr.7: 141061109 - 141072119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177897.3 NP_808565.2 6 - 7 643 75
GenBank mRNA AB114827.1 - 6 - 7 643 75
BC031982.1 - 5 - 6 497 75
BC062653.1 - 2 - 3 333 75

View Full Product Details