Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sat1
Entrez Gene ID
20229
Gene Name
spermidine/spermine N1-acetyl transferase 1
Gene Aliases
AA617398, SSAT, Sat
Chromosome Location
Chr.X: 155213126 - 155216449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291865.1 NP_001278794.1 3 - 4 484 94

View Full Product Details