Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gli3
Entrez Gene ID
14634
Gene Name
GLI-Kruppel family member GLI3
Gene Aliases
AI854843, AU023367, Bph, GLI3-190, GLI3FL, Pdn, Xt, add
Chromosome Location
Chr.13: 15463235 - 15730026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008130.2 NP_032156.2 8 - 9 1675 97
GenBank mRNA AK144851.1 - 8 - 9 1459 97
AK158261.1 - 8 - 9 1675 97
BC141135.1 - 8 - 9 1376 97
BC145445.1 - 8 - 9 1376 97
X95255.1 - 8 - 9 1415 97

View Full Product Details