Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phex
Entrez Gene ID
18675
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
Gy, HPDR, HPDR1, Hyp, PEX
Chromosome Location
Chr.X: 157162075 - 157416551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011077.2 NP_035207.1 16 - 17 2229 80
GenBank mRNA AK158481.1 - 16 - 17 2256 80
BC137925.1 - 16 - 17 1976 80
BC137926.1 - 16 - 17 1976 80
EF194891.1 - 16 - 17 1763 80
U49908.1 - 16 - 17 2221 80
U73910.1 - 16 - 17 2229 80
U75646.1 - 16 - 17 1810 80

View Full Product Details