Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phex
Entrez Gene ID
18675
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
Gy, HPDR, HPDR1, Hyp, PEX
Chromosome Location
Chr.X: 157162075 - 157416551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011077.2 NP_035207.1 18 - 19 2431 150
GenBank mRNA AK158481.1 - 18 - 19 2458 150
BC137925.1 - 18 - 19 2178 150
BC137926.1 - 18 - 19 2178 150
EF194891.1 - 18 - 19 1965 150
U49908.1 - 18 - 19 2423 150
U73910.1 - 18 - 19 2431 150
U75646.1 - 18 - 19 2012 150

View Full Product Details