Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif1ax
Entrez Gene ID
66235
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Gene Aliases
1500010B24Rik, AI426898, Eif1ay
Chromosome Location
Chr.X: 159372195 - 159385699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025437.4 NP_079713.2 1 - 2 222 69
GenBank mRNA AK005185.1 - 1 - 2 210 69
AK007301.1 - 1 - 2 173 69
AK012233.1 - 1 - 2 202 69
AK012464.1 - 1 - 2 123 69
AK012714.1 - 1 - 2 205 69
AK021283.1 - 1 - 2 203 69
AK030774.1 - - 226 69
AK031113.1 - - 234 69
AK077710.1 - 1 - 2 171 69
AK090053.1 - - 188 69
AK134827.1 - - 147 69
AK134969.1 - - 147 69
AK165574.1 - - 188 69
AK166629.1 - 1 - 2 225 69
AK166852.1 - 1 - 2 223 69
BC027284.1 - 1 - 2 159 69

View Full Product Details