Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zpr1
Entrez Gene ID
22687
Gene Name
ZPR1 zinc finger
Gene Aliases
AI303781, Zfp259, Znf259
Chromosome Location
Chr.9: 46273064 - 46282643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011752.2 NP_035882.1 11 - 12 1226 67
GenBank mRNA AB041612.1 - 11 - 12 1109 67
AK016551.1 - 5 - 6 393 67
AK044472.1 - 11 - 12 1115 67
AK167849.1 - 11 - 12 1097 67
AK168061.1 - 11 - 12 1068 67
AK194385.1 - 2 - 3 187 67
BC021397.1 - 11 - 12 1109 67

View Full Product Details