Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zpr1
Entrez Gene ID
22687
Gene Name
ZPR1 zinc finger
Gene Aliases
AI303781, Zfp259, Znf259
Chromosome Location
Chr.9: 46273064 - 46282643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011752.2 NP_035882.1 1 - 2 307 98
GenBank mRNA AB041612.1 - 1 - 2 190 98
AK044472.1 - 1 - 2 196 98
AK158995.1 - 1 - 2 268 98
AK167849.1 - 1 - 2 178 98
AK168061.1 - 1 - 2 149 98
AK170372.1 - 1 - 2 234 98
AK215085.1 - 1 - 2 78 98
BC021397.1 - 1 - 2 190 98

View Full Product Details