Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsl1d1
Entrez Gene ID
66409
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
2410005K20Rik, C76439, pBK1
Chromosome Location
Chr.16: 11193037 - 11203292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025546.2 NP_079822.1 7 - 8 883 68
GenBank mRNA AK012937.1 - 7 - 8 876 68
AK075933.1 - 7 - 8 884 68
AK136025.1 - 7 - 8 850 68
AK145699.1 - 7 - 8 897 68
AK162241.1 - 7 - 8 857 68
AK166579.1 - 7 - 8 879 68
AK168469.1 - 7 - 8 879 68
AK168717.1 - 7 - 8 872 68
AK171805.1 - 7 - 8 871 68
BC034355.1 - 7 - 8 849 68
BC069934.1 - 7 - 8 857 68
DQ445383.1 - 7 - 8 891 68

View Full Product Details