The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023348.4 | NP_075837.3 | 3 - 4 | 641 | 99 |
GenBank mRNA | AK005042.1 | - | 3 - 4 | 591 | 99 |
AK007065.1 | - | 3 - 4 | 634 | 99 | |
AK039203.1 | - | 3 - 4 | 610 | 99 | |
AK147845.1 | - | 3 - 4 | 549 | 99 | |
AY008722.1 | - | 3 - 4 | 638 | 99 | |
BC030066.1 | - | 3 - 4 | 590 | 99 |