Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfsd12
Entrez Gene ID
73822
Gene Name
major facilitator superfamily domain containing 12
Gene Aliases
C19orf28, F630110N24Rik, Wdt1
Chromosome Location
Chr.10: 81357469 - 81366225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028657.3 NP_082933.2 1 - 2 316 148
GenBank mRNA AK149678.1 - 1 - 2 322 148
AK154386.1 - 1 - 2 318 148
AK154902.1 - 1 - 2 396 148
AK170185.1 - 1 - 2 336 148
AK171084.1 - 1 - 2 318 148
BC118620.1 - 1 - 2 316 148
BC119790.1 - 1 - 2 316 148

View Full Product Details