Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Micu1
Entrez Gene ID
216001
Gene Name
mitochondrial calcium uptake 1
Gene Aliases
C730016L05Rik, Calc, Cbara1
Chromosome Location
Chr.10: 59702482 - 59864134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144822.3 NP_659071.1 5 - 6 670 107
GenBank mRNA AK049521.1 - 5 - 6 666 107
AK050113.1 - 5 - 6 657 107
AK052485.1 - 5 - 6 618 107
AK077605.1 - 5 - 6 620 107
AK085553.1 - 5 - 6 589 107
AK189367.1 - 2 - 3 93 107
AK200391.1 - 2 - 3 93 107
BC018320.1 - 5 - 6 611 107
BC023042.1 - 4 - 5 554 107
BC026566.1 - 5 - 6 621 107

View Full Product Details