The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025534.2 | NP_079810.2 | 7 - 8 | 1453 | 133 |
GenBank mRNA | AK088611.1 | - | 2 - 3 | 420 | 133 |
BC040405.1 | - | 5 - 6 | 614 | 133 | |
BC057326.1 | - | 7 - 8 | 1453 | 133 | |
BC098496.1 | - | 6 - 7 | 1380 | 133 |