Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npffr1
Entrez Gene ID
237362
Gene Name
neuropeptide FF receptor 1
Gene Aliases
Gm236, Gpr147, NPFF1, OT7T022
Chromosome Location
Chr.10: 61595484 - 61628762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177511.1 NP_001170982.1 1 - 2 336 59
GenBank mRNA AY255578.1 - 1 - 2 268 59

View Full Product Details