Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp1r14a
Entrez Gene ID
68458
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Aliases
1110001M11Rik, Cpi17
Chromosome Location
Chr.7: 29289320 - 29293390 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026731.3 NP_081007.2 2 - 3 294 79
GenBank mRNA AF352573.1 - 2 - 3 302 79
AY050672.1 - 2 - 3 293 79
BC010832.1 - 2 - 3 294 79

View Full Product Details