Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cspg4
Entrez Gene ID
121021
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
4732461B14Rik, AN2, NG2
Chromosome Location
Chr.9: 56865104 - 56899870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139001.2 NP_620570.2 8 - 9 5047 73
GenBank mRNA AF352400.1 - 8 - 9 4991 73
AK140986.1 - 3 - 4 455 73

View Full Product Details