The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199222.3 | NP_954692.2 | 12 - 13 | 1491 | 86 |
GenBank mRNA | AB211986.1 | - | 12 - 13 | 1412 | 86 |