Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grik5
Entrez Gene ID
14809
Gene Name
glutamate receptor, ionotropic, kainate 5 (gamma 2)
Gene Aliases
GluK5, GluRgamma2, KA2
Chromosome Location
Chr.7: 25009849 - 25072499 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008168.2 NP_032194.2 19 - 20 2899 87
GenBank mRNA BC052009.1 - 19 - 20 2787 87
BC058097.1 - 19 - 20 2658 87
BC110682.1 - 19 - 20 2822 87
D10011.1 - 19 - 20 2828 87

View Full Product Details