Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grik5
Entrez Gene ID
14809
Gene Name
glutamate receptor, ionotropic, kainate 5 (gamma 2)
Gene Aliases
GluK5, GluRgamma2, KA2
Chromosome Location
Chr.7: 25009849 - 25072499 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008168.2 NP_032194.2 7 - 8 1122 69
GenBank mRNA BC052009.1 - 7 - 8 1010 69
BC058097.1 - 7 - 8 881 69
BC110682.1 - 7 - 8 1045 69
D10011.1 - 7 - 8 1051 69

View Full Product Details