Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rinl
Entrez Gene ID
320435
Gene Name
Ras and Rab interactor-like
Gene Aliases
5830482F20Rik, 9930116N10Rik
Chromosome Location
Chr.7: 28788861 - 28798965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC044761.1 - 1 - 2 43 65
BC046530.1 - 1 - 2 43 65

View Full Product Details