Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrh1
Entrez Gene ID
15465
Gene Name
histamine receptor H1
Gene Aliases
Bphs, H1R, HH1R, Hir
Chromosome Location
Chr.6: 114397925 - 114483296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252643.1 NP_001239572.1 2 - 3 294 58
GenBank mRNA AK047070.1 - 2 - 3 135 58

View Full Product Details