The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163161.1 | NP_001156633.1 | 1 - 2 | 257 | 69 |
NM_010819.4 | NP_034949.3 | 1 - 2 | 257 | 69 | |
GenBank mRNA | AF061272.1 | - | 1 - 2 | 189 | 69 |
AK089500.1 | - | 1 - 2 | 248 | 69 | |
AK150926.1 | - | 1 - 2 | 163 | 69 | |
AK155919.1 | - | 1 - 2 | 255 | 69 | |
AK170692.1 | - | 1 - 2 | 195 | 69 | |
AK171409.1 | - | 1 - 2 | 195 | 69 | |
BC099471.1 | - | 1 - 2 | 167 | 69 | |
BY350724.1 | - | 1 - 2 | 260 | 69 |