The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023060.3 | NP_075547.1 | 1 - 2 | 334 | 68 |
GenBank mRNA | AF268871.1 | - | 1 - 2 | 298 | 68 |
AF283518.1 | - | 1 - 2 | 289 | 68 | |
AK028424.1 | - | 1 - 2 | 316 | 68 | |
AK087501.1 | - | 1 - 2 | 322 | 68 | |
AK147924.1 | - | 1 - 2 | 303 | 68 | |
AK171885.1 | - | 1 - 2 | 314 | 68 | |
BC024915.1 | - | 1 - 2 | 308 | 68 | |
BC099675.1 | - | 1 - 2 | 292 | 68 |