Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfpt1
Entrez Gene ID
14583
Gene Name
glutamine fructose-6-phosphate transaminase 1
Gene Aliases
2810423A18Rik, AI324119, AI449986, GFA, GFAT, GFAT1, GFAT1m, Gfpt
Chromosome Location
Chr.6: 87042846 - 87092207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013528.3 NP_038556.1 11 - 12 87
GenBank mRNA AF334736.1 - 12 - 13 87
AK019852.1 - 11 - 12 87
BC002283.1 - 8 - 9 800 87
BC010516.1 - 11 - 12 1149 87
BC050762.1 - 11 - 12 87

View Full Product Details