The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145571.2 | NP_663546.1 | 5 - 6 | 783 | 72 |
GenBank mRNA | AK028416.1 | - | 5 - 6 | 806 | 72 |
AK028695.1 | - | 3 - 4 | 516 | 72 | |
AK158228.1 | - | 5 - 6 | 767 | 72 | |
AK167515.1 | - | 5 - 6 | 778 | 72 | |
AK170691.1 | - | 5 - 6 | 778 | 72 | |
AK172205.1 | - | 5 - 6 | 786 | 72 | |
BC009149.1 | - | 5 - 6 | 783 | 72 | |
BC033463.1 | - | 5 - 6 | 737 | 72 |