The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168333.1 | NP_001161805.1 | 6 - 7 | 792 | 75 |
NM_023476.3 | NP_075965.2 | 6 - 7 | 933 | 75 | |
GenBank mRNA | AB047402.1 | - | 6 - 7 | 934 | 75 |
AB050626.1 | - | 6 - 7 | 770 | 75 | |
AK087142.1 | - | - | 793 | 75 | |
BC005738.1 | - | 6 - 7 | 760 | 75 | |
BC018539.1 | - | 6 - 7 | 777 | 75 | |
CT010275.1 | - | 5 - 6 | 551 | 75 |