The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199307.2 | NP_955011.1 | 3 - 4 | 317 | 102 |
GenBank mRNA | AK134088.1 | - | 3 - 4 | 318 | 102 |
AK160504.1 | - | 2 - 3 | 170 | 102 | |
BC060648.1 | - | 3 - 4 | 257 | 102 |