Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alms1
Entrez Gene ID
236266
Gene Name
Alstrom syndrome 1
Gene Aliases
bbb
Chromosome Location
Chr.6: 85587499 - 85726611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145223.2 NP_660258.2 15 - 16 7778 136
GenBank mRNA AF425257.1 - 15 - 16 7779 136

View Full Product Details