The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198884.1 | NP_942585.1 | 19 - 20 | 2855 | 89 |
GenBank mRNA | AB114826.1 | - | 19 - 20 | 2855 | 89 |
AK082874.1 | - | 5 - 6 | 574 | 89 | |
AK143898.1 | - | 19 - 20 | 2771 | 89 | |
BC139798.1 | - | 19 - 20 | 2869 | 89 |