Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdtc1
Entrez Gene ID
230796
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
Gm695, adipose, adp
Chromosome Location
Chr.4: 133292466 - 133353476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199306.4 NP_955010.1 12 - 13 1653 83
GenBank mRNA AK131700.1 - 14 - 15 1809 83
AK172431.1 - 12 - 13 1699 83
BC034833.1 - 5 - 6 576 83
BC048824.1 - 12 - 13 1447 83
BC057107.1 - 11 - 12 1296 83

View Full Product Details