Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1bg
Entrez Gene ID
117586
Gene Name
alpha-1-B glycoprotein
Gene Aliases
C44
Chromosome Location
Chr.15: 60917589 - 60921270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081067.1 NP_001074536.1 5 - 6 960 62
GenBank mRNA BC080745.1 - - 718 62
DQ520794.1 - 5 - 6 956 62

View Full Product Details