Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sntb1
Entrez Gene ID
20649
Gene Name
syntrophin, basic 1
Gene Aliases
-
Chromosome Location
Chr.15: 55638505 - 55907305 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016667.3 NP_057876.1 2 - 3 1143 126
GenBank mRNA U89997.1 - 2 - 3 1148 126

View Full Product Details