Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl13
Entrez Gene ID
68537
Gene Name
mitochondrial ribosomal protein L13
Gene Aliases
1110002D09Rik
Chromosome Location
Chr.15: 55534095 - 55557312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026759.3 NP_081035.1 2 - 3 175 144
GenBank mRNA AB049641.1 - 2 - 3 153 144
AK003283.2 - 2 - 3 176 144
BC055470.1 - 2 - 3 139 144
BC060968.1 - 2 - 3 181 144

View Full Product Details