Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asah1
Entrez Gene ID
11886
Gene Name
N-acylsphingosine amidohydrolase 1
Gene Aliases
2310081N20Rik, AC, Asah
Chromosome Location
Chr.8: 41340643 - 41374697 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019734.2 NP_062708.1 2 - 3 230 78
GenBank mRNA AF157500.1 - 2 - 3 160 78
AF352179.1 - 2 - 3 172 78
AK075666.1 - 2 - 3 145 78
AK075919.1 - 2 - 3 184 78
AK077721.1 - 2 - 3 183 78
AK090314.1 - 2 - 3 180 78
AK151208.1 - 2 - 3 231 78
AK151478.1 - 2 - 3 221 78
AK152305.1 - 2 - 3 182 78
AK152920.1 - 2 - 3 190 78
AK153101.1 - 2 - 3 190 78
AK153297.1 - 2 - 3 144 78
AK159556.1 - 2 - 3 152 78
AK166757.1 - 2 - 3 162 78
BC003204.1 - 2 - 3 150 78
BY160973.1 - 2 - 3 231 78

View Full Product Details