The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253877.1 | NP_001240806.1 | 5 - 6 | 1030 | 86 |
NM_001253878.1 | NP_001240807.1 | 5 - 6 | 749 | 86 | |
NM_001253879.1 | NP_001240808.1 | 4 - 5 | 672 | 86 | |
NM_001253880.1 | NP_001240809.1 | 2 - 3 | 387 | 86 | |
NM_013827.3 | NP_038855.2 | 4 - 5 | 668 | 86 | |
NR_045608.1 | 6 - 7 | 736 | 86 | ||
GenBank mRNA | AB221622.1 | - | 1 - 2 | 75 | 86 |
AK030752.1 | - | 4 - 5 | 618 | 86 | |
AK031409.1 | - | 6 - 7 | 737 | 86 | |
AK032798.1 | - | 5 - 6 | 991 | 86 | |
AK032819.1 | - | 5 - 6 | 992 | 86 | |
AK137539.1 | - | 6 - 7 | 977 | 86 | |
AK139685.1 | - | 5 - 6 | 750 | 86 | |
BC024889.1 | - | 4 - 5 | 640 | 86 | |
BC029076.1 | - | 4 - 5 | 579 | 86 | |
BC050826.1 | - | 4 - 5 | 622 | 86 | |
BC057163.1 | - | 4 - 5 | 465 | 86 | |
BC071223.1 | - | 2 - 3 | 140 | 86 | |
BC092237.1 | - | 4 - 5 | 600 | 86 | |
BC100340.1 | - | 2 - 3 | 318 | 86 | |
S78454.1 | - | 1 - 2 | 75 | 86 |