The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005863.2 | NP_001005863.1 | 4 - 5 | 2804 | 83 |
NM_001005864.3 | NP_001005864.1 | 2 - 3 | 410 | 83 | |
NM_001286413.1 | NP_001273342.1 | 4 - 5 | 2686 | 83 | |
GenBank mRNA | AK043214.1 | - | 2 - 3 | 397 | 83 |
AY626782.1 | - | 2 - 3 | 408 | 83 | |
AY626783.1 | - | 3 - 4 | 2626 | 83 | |
BB647317.1 | - | 2 - 3 | 410 | 83 | |
BC041777.1 | - | 3 - 4 | 1469 | 83 | |
BC042206.1 | - | 3 - 4 | 1469 | 83 | |
BC043321.1 | - | 2 - 3 | 374 | 83 | |
BC089009.1 | - | 4 - 5 | 2686 | 83 |