Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnrnpd
Entrez Gene ID
11991
Gene Name
heterogeneous nuclear ribonucleoprotein D
Gene Aliases
Auf1, Hnrpd
Chromosome Location
Chr.5: 99955935 - 99979015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077267.2 NP_001070735.1 5 - 6 1114 60
NM_007516.3 NP_031542.2 6 - 7 1171 60
GenBank mRNA AK077409.1 - 6 - 7 1159 60
BC049098.1 - 6 - 7 1171 60
CF751545.1 - 3 - 4 322 60

View Full Product Details